Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080743.1 | NP_001074212.1 | 3 | 802 | ||
NM_019497.2 | NP_062370.2 | 3 | 802 | |||
GenBank mRNA | AF040745.1 | - | 3 | 228 | ||
AK020792.1 | - | 3 | 802 | |||
AK078475.1 | - | 3 | 674 | |||
AK132685.1 | - | 3 | 803 | |||
BC150691.1 | - | 3 | 326 | 9087602 |