Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021332.2 | NP_067307.2 | 11 | 1147 | ||
GenBank mRNA | AJ001692.1 | - | 11 | 1137 | ||
AY293742.1 | - | 11 | 1137 | |||
BC103624.1 | - | 11 | 1136 | |||
BC105649.1 | - | 11 | 1136 | |||
BC105650.1 | - | 11 | 1136 | |||
BC139464.1 | - | 11 | 1174 | 8862486 | ||
BC139466.1 | - | 11 | 1174 | 8862488 | ||
BC145626.1 | - | 11 | 1174 | 9054194 | ||
DQ093397.1 | - | 11 | 1147 | |||
DQ093398.1 | - | 11 | 1147 |
Set Membership: |
G-protein coupled receptor |