Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021355.3 | NP_067330.1 | 2 | 828 | ||
GenBank mRNA | AK032302.1 | - | 2 | 829 | ||
AK053317.1 | - | 2 | 829 | |||
AK054183.1 | - | 2 | 885 | |||
AK080601.1 | - | 2 | 846 | |||
AK082593.1 | - | 2 | 845 | |||
BC052673.1 | - | 2 | 837 | |||
BC064779.1 | - | 2 | 801 | 30058603 | ||
X94998.1 | - | 2 | 800 |