Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HSPA8 SIRNA Assays ›
Gene Symbol:
Hspa8
Gene Name:
heat shock protein 8
Gene Aliases:
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
Chromosome Location:
Chr.9: 40801273 - 40805199 on Build GRCm38
UniGene ID:
Mm.290774
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
159532
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hspa8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031165.4
NP_112442.2
5
660
GenBank mRNA
AK004608.1
-
4
949
AK035286.1
-
5
658
AK075935.1
-
5
657
AK081701.1
-
5
659
AK098074.1
-
5
657
AK145416.1
-
5
580
AK145579.1
-
5
654
AK146547.1
-
5
661
AK146708.1
-
5
655
AK146985.1
-
5
658
AK147864.1
-
5
658
AK149954.1
-
5
655
AK150474.1
-
5
652
AK150498.1
-
5
652
AK150701.1
-
5
662
AK150958.1
-
5
655
AK151041.1
-
5
658
AK151065.1
-
5
658
AK151127.1
-
5
661
AK151287.1
-
5
658
AK151435.1
-
5
658
AK151516.1
-
5
664
AK151537.1
-
5
655
AK151741.1
-
5
658
AK151775.1
-
5
660
AK151808.1
-
5
661
AK151865.1
-
5
658
AK151892.1
-
5
660
AK151948.1
-
5
661
AK151997.1
-
5
658
AK152315.1
-
5
661
AK152598.1
-
5
658
AK152697.1
-
5
658
AK152703.1
-
5
658
AK152768.1
-
5
661
AK152803.1
-
5
661
AK153032.1
-
5
660
AK153834.1
-
5
658
AK157831.1
-
5
658
AK159479.1
-
5
658
AK162775.1
-
5
658
AK163987.1
-
5
658
AK164000.1
-
5
658
AK166545.1
-
5
660
AK166643.1
-
5
672
AK166721.1
-
5
655
AK166767.1
-
5
658
AK166776.1
-
5
661
AK166808.1
-
5
658
AK166830.1
-
5
656
AK166846.1
-
5
658
AK166861.1
-
5
661
AK166873.1
-
5
658
AK166908.1
-
5
661
AK166910.1
-
5
658
AK166913.1
-
5
658
AK166933.1
-
5
658
AK167043.1
-
5
660
AK167121.1
-
5
661
AK167122.1
-
5
658
AK167134.1
-
5
656
AK167163.1
-
5
660
AK167218.1
-
5
658
AK167229.1
-
5
659
AK167254.1
-
5
659
AK167414.1
-
5
658
AK167845.1
-
5
661
AK167910.1
-
5
661
AK168435.1
-
5
661
AK168492.1
-
5
660
AK168511.1
-
3
196
AK168519.1
-
5
658
AK168542.1
-
5
660
AK168577.1
-
5
658
AK168625.1
-
5
658
AK168654.1
-
5
658
AK168711.1
-
5
658
AK168750.1
-
5
661
AK168776.1
-
5
661
AK168887.1
-
5
660
AK168934.1
-
5
661
AK169093.1
-
5
658
AK169179.1
-
5
658
AK169236.1
-
5
658
AK169293.1
-
5
672
AK169329.1
-
5
661
AK169594.1
-
5
657
AK171434.1
-
5
659
AK178143.1
-
2
139
AK184919.1
-
2
139
AK190509.1
-
2
139
AK192182.1
-
2
140
AK192272.1
-
2
139
AK193850.1
-
2
139
AK193882.1
-
2
139
AK195240.1
-
2
139
AK198438.1
-
2
139
AK199049.1
-
3
366
AK199142.1
-
2
139
AK199178.1
-
2
139
AK199354.1
-
2
139
AK199410.1
-
2
172
AK199504.1
-
2
139
AK200094.1
-
2
139
AK200309.1
-
2
139
AK200579.1
-
2
139
AK201767.1
-
2
139
AK202859.1
-
2
139
AK203911.1
-
2
139
AK204890.1
-
2
139
AK205187.1
-
2
139
AK206134.1
-
2
139
AK206953.1
-
2
139
AK209289.1
-
2
139
AK210012.1
-
3
205
AK211519.1
-
2
139
AK212081.1
-
3
203
AK212914.1
-
2
159
AK214003.1
-
3
186
AK216159.1
-
2
139
AK216732.1
-
2
139
BC006722.1
-
5
682
3711399
BC066191.1
-
5
614
30467052
BC085486.1
-
5
646
30540914
BC089322.1
-
5
639
30538640
BC089457.1
-
5
617
30628505
BC094900.1
-
5
574
30063483
BC106169.1
-
3
338
BC106193.1
-
5
642
6329283
M19141.1
-
5
644
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
protein refolding
negative regulation of transcription, DNA-templated
ATP metabolic process
positive regulation of mRNA splicing, via spliceosome
chaperone mediated protein folding requiring cofactor
regulation of cell cycle
chaperone-mediated protein folding
clathrin coat disassembly
negative regulation of fibril organization
nucleotide binding
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
MHC class II protein complex binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
ADP binding
poly(A) RNA binding
unfolded protein binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0