Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133975.4 | NP_598736.4 | 20 | 3086 | ||
GenBank mRNA | AK033327.1 | - | 3 | 401 | ||
BC034113.1 | - | 4 | 492 |