Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOPEY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320714.1 | NP_001307643.1 | 36 | 6810 | ||
NM_005128.3 | NP_005119.2 | 36 | 6701 | |||
XM_017028509.1 | XP_016883998.1 | 33 | 6204 | |||
GenBank mRNA | AB023150.2 | - | 33 | 6170 | ||
AJ237839.2 | - | 36 | 6707 | |||
AK026478.1 | - | 15 | 1637 | |||
BC012791.2 | - | 18 | 2322 |