Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039515.1 | NP_001034604.1 | 2 | 1494 | ||
NM_007487.3 | NP_031513.1 | 2 | 1458 | |||
GenBank mRNA | AK048606.1 | - | 2 | 1459 | ||
BC029234.1 | - | 2 | 1418 | 3979729 |