Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007496.2 | NP_031522.2 | 2 | 2447 | ||
GenBank mRNA | D26046.1 | - | 2 | 2444 |