Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007499.2 | NP_031525.2 | 10 | 1684 | ||
GenBank mRNA | BC138525.1 | - | 9 | 1586 | 8861545 | |
U43678.1 | - | 9 | 1586 |