Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301424.1 | NP_001288353.1 | 6 | 868 | ||
NM_007513.4 | NP_031539.3 | 6 | 984 | |||
GenBank mRNA | AK139342.1 | - | 6 | 960 | ||
AK147991.1 | - | 6 | 893 | |||
AK163202.1 | - | 6 | 868 | |||
BC068234.1 | - | 6 | 875 | |||
BC145779.1 | - | 5 | 789 | 40131082 | ||
BC145781.1 | - | 5 | 789 | 40131084 | ||
JN226407.1 | - | 4 | 704 | |||
JN226408.1 | - | 4 | 704 | |||
M26687.1 | - | 6 | 902 |