Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005167.5 | NP_005158.5 | 10 | 1663 | ||
GenBank mRNA | AK093270.1 | - | 10 | 1643 | ||
AL133644.1 | - | 11 | 2427 | |||
BC011803.2 | - | 10 | 1587 | |||
BC064628.1 | - | 10 | 1445 | |||
BC073828.1 | - | 10 | 1655 | 5244363 |