Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048139.1 | NP_001041604.1 | 2 | 4071 | ||
NM_001048141.1 | NP_001041606.1 | 2 | 3911 | |||
NM_001048142.1 | NP_001041607.1 | 2 | 3919 | |||
NM_001285416.1 | NP_001272345.1 | 2 | 3988 | |||
NM_001285417.1 | NP_001272346.1 | 2 | 3785 | |||
NM_001285418.1 | NP_001272347.1 | 2 | 3776 | |||
NM_001285419.1 | NP_001272348.1 | 2 | 3765 | |||
NM_001285420.1 | NP_001272349.1 | 2 | 3653 | |||
NM_001285421.1 | NP_001272350.1 | 2 | 3851 | |||
NM_001285422.1 | NP_001272351.1 | 1 | 4033 | |||
NM_007540.4 | NP_031566.4 | 2 | 4212 | |||
GenBank mRNA | AY057908.1 | - | 2 | 4208 | ||
AY057909.1 | - | 2 | 3772 | |||
AY057910.1 | - | 2 | 3984 | |||
AY057911.1 | - | 2 | 4067 | |||
AY057912.1 | - | 2 | 3699 | |||
AY057913.1 | - | 2 | 3907 | |||
AY057914.1 | - | 2 | 3915 | |||
AY231131.1 | - | 2 | 3721 | |||
AY231132.1 | - | 3 | 3796 | |||
EF125669.1 | - | 2 | 4212 | |||
EF125670.1 | - | 2 | 3777 | |||
EF125671.1 | - | 2 | 3989 | |||
EF125672.1 | - | 2 | 4073 | |||
EF125673.1 | - | 2 | 3911 | |||
EF125674.1 | - | 2 | 3919 | |||
EF125681.1 | - | 2 | 3765 | |||
EF125682.1 | - | 2 | 3653 | |||
EF125683.1 | - | 2 | 3785 | |||
EF125684.1 | - | 2 | 3851 | |||
EF125685.1 | - | 1 | 4030 |