Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005195.3 | NP_005186.2 | 1 | 1184 | ||
GenBank mRNA | BC094715.1 | - | 2 | 1396 | ||
M83667.1 | - | 1 | 1219 |