Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005218.3 | NP_005209.1 | 2 | 282 | ||
GenBank mRNA | BC033298.1 | - | 2 | 192 | 4775720 | |
BC047677.1 | - | 2 | 199 | 5759899 | ||
U73945.1 | - | 2 | 203 | |||
X92744.1 | - | 2 | 199 | |||
Z50788.1 | - | 1 | 36 |