Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320866.1 | NP_001307795.1 | 19 | 2355 | ||
NM_001721.6 | NP_001712.1 | 19 | 2296 | |||
NM_203281.2 | NP_975010.1 | 19 | 2358 | |||
GenBank mRNA | AF045459.1 | - | 19 | 2369 | ||
AK291824.1 | - | 19 | 2328 | |||
BC016652.2 | - | 19 | 2313 | 3919253 | ||
X83107.1 | - | 19 | 2218 |