Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007649.4 | NP_031675.1 | 2 | 204 | ||
GenBank mRNA | AB196820.1 | - | 2 | 165 | ||
AB196821.1 | - | 2 | 165 | |||
AB196822.1 | - | 2 | 165 | |||
AB196823.1 | - | 2 | 165 | |||
AK007741.1 | - | 2 | 205 | |||
AK084984.1 | - | 2 | 233 | |||
AK088027.1 | - | 2 | 251 | |||
AK146173.1 | - | 2 | 251 | |||
AK151601.1 | - | 2 | 195 | |||
BC060977.1 | - | 2 | 232 | 30296173 | ||
X17501.1 | - | 2 | 215 | |||
X53526.1 | - | 2 | 215 |