Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007650.3 | NP_031676.3 | 6, 7 | 1218 | ||
GenBank mRNA | AK089076.1 | - | 6, 7 | 1166 | ||
AK143736.1 | - | 6, 7 | 1194 | |||
BC011258.1 | - | 6, 7 | 1218 | 4188683 | ||
M15177.1 | - | 6, 7 | 1096 | |||
U65961.1 | - | 6, 7 | 1096 |