Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007672.2 | NP_031698.2 | 3 | 478 | ||
GenBank mRNA | AK148057.1 | - | 3 | 462 | ||
AK171706.1 | - | 3 | 433 | |||
BC117530.1 | - | 3 | 367 | 40106297 | ||
BC118511.1 | - | 3 | 367 | 40106298 | ||
BY713743.1 | - | 3 | 478 | |||
U88588.1 | - | 4 | 371 |