Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007672.2 | NP_031698.2 | 5 | 1987 | ||
GenBank mRNA | AK148057.1 | - | 5 | 1971 | ||
AK171706.1 | - | 5 | 1942 | |||
CF616011.1 | - | 1 | 222 | |||
DT902919.1 | - | 1 | 534 | |||
U88588.1 | - | 6 | 1881 |