Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007703.2 | NP_031729.1 | 4 | 988 | ||
GenBank mRNA | AK162271.1 | - | 1 | 338 | ||
BC016468.1 | - | 4 | 983 | 4196583 | ||
U97107.1 | - | 4 | 988 |