Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF033566.1 | - | 3 | 370 | ||
AK013974.2 | - | 2 | 216 | |||
AK051099.1 | - | 1 | 2476 | |||
AY653308.1 | - | 3 | 351 | |||
BC012675.1 | - | 3 | 392 | 3982920 | ||
CT010293.1 | - | 2 | 292 | |||
U94846.1 | - | 3 | 389 |