Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007808.4 | NP_031834.1 | 3 | 378 | ||
GenBank mRNA | AK002732.1 | - | 3 | 380 | ||
AK003168.1 | - | 3 | 407 | |||
AK005336.1 | - | 3 | 450 | |||
AK088098.1 | - | 3 | 379 | |||
AK143887.1 | - | 3 | 379 | |||
AK168017.1 | - | 3 | 379 | |||
BC034363.1 | - | 3 | 362 | 2650742 | ||
BC092213.1 | - | 2 | 327 | 6845432 | ||
BC094054.1 | - | 3 | 367 | 2865963 |