Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007843.3 | NP_031869.1 | 2 | 347 | ||
GenBank mRNA | AI956736.1 | - | 2 | 413 | ||
AK142544.1 | - | 2 | 328 | |||
BC024380.1 | - | 2 | 303 | 4975536 |