Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dffb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007859.4 | NP_031885.3 | 3 | 484 | ||
GenBank mRNA | AB009377.1 | - | 3 | 392 | ||
AK036094.1 | - | 3 | 461 | |||
AK077619.1 | - | 3 | 530 | |||
AK089267.1 | - | 3 | 461 | |||
BC053052.1 | - | 3 | 414 | 6403143 |