Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301211.1 | NP_001288140.1 | 3 | 656 | ||
NM_009677.6 | NP_033807.2 | 3 | 656 | |||
GenBank mRNA | AK036387.1 | - | 3 | 599 | ||
AK145821.1 | - | 3 | 578 | |||
BC006691.1 | - | 3 | 333 | |||
BC052703.1 | - | 3 | 529 | 5698075 | ||
BC054535.1 | - | 3 | 529 | 5698075 | ||
X54424.1 | - | 3 | 578 |