Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009700.2 | NP_033830.2 | 4 | 693 | ||
GenBank mRNA | AF469168.1 | - | 4 | 767 | ||
AF469169.1 | - | 4 | 949 | |||
AK039777.1 | - | 4 | 696 | |||
AK044074.1 | - | 4 | 632 | |||
AK045357.1 | - | 3 | 344 | |||
AY484964.1 | - | 4 | 699 | |||
AY484965.1 | - | 5 | 883 | |||
AY484966.1 | - | 5 | 829 | |||
AY728043.1 | - | 4 | 713 | |||
AY728044.1 | - | 5 | 897 | |||
AY728045.1 | - | 4 | 825 | |||
AY803942.1 | - | 5 | 804 | |||
AY803943.1 | - | 6 | 934 | |||
AY803944.1 | - | 7 | 1118 | |||
BC024526.1 | - | 4 | 639 | 4985265 | ||
CT010362.1 | - | 3 | 640 | |||
U33012.1 | - | 3 | 655 | |||
U48397.1 | - | 3 | 775 | |||
U48398.1 | - | 3 | 646 | |||
U48399.1 | - | 5 | 757 | |||
U88623.1 | - | 4 | 694 |