Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009700.2 | NP_033830.2 | 5 | 1151 | ||
GenBank mRNA | AK039777.1 | - | 5 | 1154 | ||
AK045357.1 | - | 4 | 802 | |||
BB195919.1 | - | 1 | 257 | |||
BC024526.1 | - | 5 | 1097 | 4985265 | ||
U88623.1 | - | 5 | 1152 |