Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290788.1 | NP_001277717.1 | 6 | 527 | ||
NM_009702.3 | NP_033832.2 | 6 | 527 | |||
GenBank mRNA | AK016322.1 | - | 6 | 526 | ||
AK020468.1 | - | 6 | 527 | |||
AK077886.1 | - | 6 | 517 | |||
AK082873.1 | - | 6 | 466 | |||
AK122310.1 | - | 6 | 515 | |||
AK138700.1 | - | 6 | 583 | |||
AK145547.1 | - | 6 | 512 | |||
AK151716.1 | - | 6 | 498 | |||
BC042479.1 | - | 6 | 507 | 5363276 | ||
U90333.1 | - | 6 | 375 |