Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290788.1 | NP_001277717.1 | 21, 22 | 2570 | ||
NM_009702.3 | NP_033832.2 | 21, 22 | 2570 | |||
GenBank mRNA | AK082873.1 | - | 21, 22 | 2512 | ||
AK122310.1 | - | 21, 22 | 2558 | |||
AK145547.1 | - | 21, 22 | 2555 | |||
BC042479.1 | - | 21, 22 | 2550 | 5363276 |