Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009703.2 | NP_033833.1 | 16 | 2064 | ||
GenBank mRNA | AK034861.1 | - | 17 | 2171 | ||
AK085823.1 | - | 16 | 2032 | |||
BC004757.1 | - | 16 | 2020 | 3154293 | ||
D00024.1 | - | 11 | 1426 |