Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009731.2 | NP_033861.2 | 10 | 1199 | ||
GenBank mRNA | AK002705.1 | - | 10 | 1201 | ||
AK147775.1 | - | 10 | 1228 | |||
BC086929.1 | - | 10 | 1172 | 6775941 | ||
J05663.1 | - | 10 | 1171 |