Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009756.2 | NP_033886.2 | 2 | 620 | ||
GenBank mRNA | AF101033.1 | - | 2 | 586 | ||
BC145044.1 | - | 2 | 620 | 9053592 | ||
BC145983.1 | - | 2 | 620 | 40131286 |