Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnip3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009761.3 | NP_033891.1 | 6 | 751 | ||
GenBank mRNA | AF067395.1 | - | 6 | 757 | ||
AK004667.1 | - | 6 | 698 | |||
AK007920.1 | - | 6 | 734 | |||
AK013467.1 | - | 6 | 733 | |||
AK018668.1 | - | 6 | 703 | |||
AK139950.1 | - | 6 | 695 | |||
AK155040.1 | - | 6 | 722 | |||
AK168482.1 | - | 6 | 752 | |||
AK169391.1 | - | 6 | 752 | |||
AY057069.1 | - | 7 | 772 | |||
BC049081.1 | - | 6 | 664 | |||
BC085237.1 | - | 6 | 701 | 5362586 |