Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009764.3 | NP_033894.3 | 4 | 373 | ||
GenBank mRNA | AB221611.1 | - | 3 | 167 | ||
AK086173.1 | - | 4 | 375 | |||
AK144702.1 | - | 4 | 292 | |||
BB662609.1 | - | 4 | 375 | |||
BC068303.1 | - | 4 | 209 | 30431022 | ||
U31625.1 | - | 4 | 230 | |||
U32446.1 | - | 4 | 283 | |||
U35641.1 | - | 4 | 266 | |||
U36475.1 | - | 3 | 193 | |||
U68174.1 | - | 4 | 277 |