Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009796.2 | NP_033926.1 | 3 | 512 | ||
GenBank mRNA | AB028640.1 | - | 3 | 512 | ||
AJ012475.1 | - | 3 | 325 | |||
AK154277.1 | - | 3 | 598 | |||
AK169334.1 | - | 3 | 557 | |||
BC095980.1 | - | 3 | 529 |