Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009800.4 | NP_033930.1 | 3 | 542 | ||
GenBank mRNA | AB079601.1 | - | 3 | 321 | ||
AF050105.1 | - | 1 | 95 | |||
AK013465.1 | - | 3 | 543 | |||
AK140073.1 | - | 3 | 643 | |||
AY075026.1 | - | 3 | 543 | |||
BC019393.1 | - | 3 | 527 | 3968862 |