Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009801.4 | NP_033931.4 | 6, 7 | 995 | ||
GenBank mRNA | AK002333.1 | - | 6, 7 | 730 | ||
AK002498.1 | - | 5, 6 | 464 | |||
AK146015.1 | - | 6, 7 | 732 | |||
BC044806.1 | - | 6, 7 | 716 | |||
BC055291.1 | - | 6, 7 | 715 | 6479187 | ||
K00811.1 | - | 6, 7 | 696 |