Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009855.2 | NP_033985.3 | 2 | 609 | ||
GenBank mRNA | AF065893.1 | - | 2 | 418 | ||
AF065894.1 | - | 2 | 418 | |||
AF065895.1 | - | 2 | 418 | |||
AF065896.1 | - | 2 | 418 | |||
AJ278965.1 | - | 2 | 361 | |||
AK154329.1 | - | 3 | 718 | |||
AY278186.1 | - | 2 | 409 | |||
BC131959.1 | - | 2 | 429 | 40130236 | ||
BC145843.1 | - | 2 | 429 | 40131146 | ||
D16220.1 | - | 2 | 424 | |||
X60958.1 | - | 2 | 609 |