Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cideb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009894.3 | NP_034024.2 | 5 | 907 | ||
GenBank mRNA | AF041377.1 | - | 5 | 894 | ||
AK003968.1 | - | 5 | 942 | |||
BC012664.1 | - | 5 | 871 | 4218866 | ||
BC046340.1 | - | 5 | 901 | 6478230 |