Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009904.2 | NP_034034.2 | 11 | 1406 | ||
GenBank mRNA | AK019534.1 | - | 11 | 1320 | ||
AK034525.1 | - | 11 | 1307 | |||
BC050767.1 | - | 11 | 1406 | 6771060 | ||
D14117.1 | - | 11 | 1419 | |||
D86323.1 | - | 11 | 1419 | |||
U08373.1 | - | 11 | 1419 |