Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036658.1 | - | 2 | 2051 | ||
AK037898.1 | - | 2 | 2161 | |||
AK134109.1 | - | 2 | 2087 | |||
BC024052.1 | - | 2 | 2016 | 5098695 | ||
X86405.1 | - | 2 | 2004 | |||
X93168.1 | - | 2 | 2055 |