Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009931.2 | NP_034061.2 | 14, 15 | 1031 | ||
GenBank mRNA | AK142097.1 | - | 14, 15 | 943 | ||
AK146487.1 | - | 14, 15 | 980 | |||
AK147284.1 | - | 14, 15 | 953 | |||
AK147355.1 | - | 14, 15 | 956 | |||
AK147661.1 | - | 14, 15 | 997 | |||
AK159130.1 | - | 14, 15 | 980 | |||
BC072650.1 | - | 14, 15 | 894 | 6852661 | ||
J04694.1 | - | 14, 15 | 935 | |||
X06777.1 | - | 14, 15 | 954 |