Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LMO7 SIRNA Assays ›
Gene Symbol:
LMO7
Gene Name:
LIM domain 7
Gene Aliases:
FBX20, FBXO20, LOMP
Chromosome Location:
Chr.13: 75620434 - 75859870 on Build GRCh38
UniGene ID:
Hs.207631
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
16134
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LMO7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306080.1
NP_001293009.1
15, 16
3327
NM_005358.5
NP_005349.3
14, 15
3371
NM_015842.2
NP_056667.2
12, 13
3018
XM_011535075.2
XP_011533377.1
15, 16
3092
XM_011535078.1
XP_011533380.1
15, 16
3092
XM_011535080.2
XP_011533382.1
15, 16
3092
XM_011535082.1
XP_011533384.1
15, 16
3092
XM_011535086.1
XP_011533388.1
14, 15
2888
XM_011535094.1
XP_011533396.1
9, 10
1715
XM_011535095.1
XP_011533397.1
12, 13
1886
XM_017020574.1
XP_016876063.1
15, 16
3092
XM_017020575.1
XP_016876064.1
15, 16
3092
XM_017020576.1
XP_016876065.1
15, 16
3092
XM_017020577.1
XP_016876066.1
15, 16
3092
XM_017020578.1
XP_016876067.1
15, 16
3092
XM_017020579.1
XP_016876068.1
15, 16
3092
XM_017020580.1
XP_016876069.1
14, 15
2888
XM_017020581.1
XP_016876070.1
14, 15
3014
XM_017020582.1
XP_016876071.1
14, 15
2888
XM_017020583.1
XP_016876072.1
14, 15
2888
XM_017020584.1
XP_016876073.1
13, 14
2810
XM_017020585.1
XP_016876074.1
13, 14
2606
XM_017020586.1
XP_016876075.1
13, 14
2606
XM_017020587.1
XP_016876076.1
13, 14
2606
XM_017020588.1
XP_016876077.1
13, 14
2606
XM_017020589.1
XP_016876078.1
13, 14
2810
XM_017020590.1
XP_016876079.1
13, 14
2606
XM_017020591.1
XP_016876080.1
14, 15
3175
XM_017020592.1
XP_016876081.1
14, 15
3058
XM_017020593.1
XP_016876082.1
13, 14
2971
XM_017020594.1
XP_016876083.1
13, 14
2854
XM_017020595.1
XP_016876084.1
13, 14
2971
XM_017020596.1
XP_016876085.1
12, 13
1886
XM_017020597.1
XP_016876086.1
11, 12
1856
XM_017020598.1
XP_016876087.1
13, 14
2971
XM_017020599.1
XP_016876088.1
12, 13
2689
XM_017020600.1
XP_016876089.1
12, 13
1886
XM_017020601.1
XP_016876090.1
12, 13
2689
XM_017020602.1
XP_016876091.1
12, 13
2572
XM_017020603.1
XP_016876092.1
12, 13
2689
XM_017020604.1
XP_016876093.1
11, 12
1969
GenBank mRNA
AB020665.2
-
12, 13
2948
AF144237.1
-
3, 4
831
AF330045.1
-
14, 15
3370
AK055465.1
-
7, 8
1983
AK092052.1
-
13, 14
2290
AK304051.1
-
10, 11
1828
FJ711162.1
-
15, 16
2957
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of signaling
regulation of cell adhesion
ubiquitin-protein transferase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0