Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009968.3 | NP_034098.1 | 4 | 923 | ||
GenBank mRNA | AK098082.1 | - | 4 | 407 | ||
AK133820.1 | - | 4 | 924 | |||
AK134281.1 | - | 4 | 408 | |||
AK208863.1 | - | 1 | 26 | |||
BC003800.1 | - | 4 | 415 | 3492859 | ||
BC043076.1 | - | 4 | 384 | 6404996 | ||
S70056.1 | - | 4 | 435 |