Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSN2 SIRNA Assays ›
Gene Symbol:
Csn2
Gene Name:
casein beta
Gene Aliases:
Csnb
Chromosome Location:
Chr.5: 87692619 - 87699429 on Build GRCm38
UniGene ID:
Mm.268737
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
161364
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Csn2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286020.1
NP_001272949.1
9
835
NM_001286021.1
NP_001272950.1
9
850
NM_001286022.1
NP_001272951.1
8
811
NM_001286023.1
NP_001272952.1
7
793
NM_001286024.1
NP_001272953.1
7
790
NM_009972.2
NP_034102.1
9
838
GenBank mRNA
AK021324.1
-
9
832
AK021328.1
-
9
832
AK052803.1
-
9
835
AK052805.1
-
9
835
AK052822.1
-
9
832
AK085678.1
-
9
832
AK085694.1
-
9
835
AK085698.1
-
9
832
AK085729.1
-
9
835
AK085756.1
-
9
838
AK142616.1
-
9
835
AK142620.1
-
9
835
AK142631.1
-
9
835
AK142635.1
-
9
835
AK164747.1
-
9
835
AK164754.1
-
2
3663
AK164761.1
-
9
835
AK164791.1
-
9
835
AK164792.1
-
9
832
AK164793.1
-
9
832
AK164794.1
-
9
835
BC003863.1
-
4
660
BC003976.1
-
5
684
BC013332.1
-
9
835
3670912
BC019114.1
-
9
816
3483918
BC019189.1
-
8
828
5028194
BC020356.1
-
9
815
BC020383.1
-
9
815
BC021153.1
-
9
833
3670372
BC050270.1
-
9
815
BC057671.1
-
6
2627
BC080709.1
-
9
819
3671502
BC092098.1
-
10
723
X04490.1
-
9
834
Back To Top
More Information
Panther Classification:
Molecular Function -
storage protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
calcium ion transport
lactation
negative regulation of catalytic activity
negative regulation of lactation
enzyme inhibitor activity
transporter activity
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0