Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010019.3 | NP_034149.2 | 10 | 1002 | ||
GenBank mRNA | AB018002.1 | - | 10 | 992 | ||
AF052942.1 | - | 8 | 682 | |||
AK004350.1 | - | 10 | 964 | |||
BC022165.1 | - | 10 | 971 | 4016911 | ||
BX527735.1 | - | 6 | 386 |