Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010024.3 | NP_034154.2 | 7 | 1823 | ||
GenBank mRNA | AK181545.1 | - | 1 | 15 | ||
AK188002.1 | - | 1 | 15 | |||
AK189987.1 | - | 1 | 15 | |||
AK191176.1 | - | 1 | 15 | |||
AK192452.1 | - | 1 | 15 | |||
AK192591.1 | - | 1 | 15 | |||
AK193231.1 | - | 1 | 15 | |||
AK195879.1 | - | 1 | 15 | |||
AK196586.1 | - | 1 | 15 | |||
AK199567.1 | - | 1 | 15 | |||
AK205371.1 | - | 1 | 15 | |||
AK206972.1 | - | 1 | 15 | |||
AK207088.1 | - | 1 | 15 | |||
AK211308.1 | - | 1 | 15 | |||
AK213122.1 | - | 1 | 15 | |||
AK213807.1 | - | 1 | 15 | |||
AK214661.1 | - | 1 | 15 | |||
AK215261.1 | - | 1 | 15 | |||
AK216173.1 | - | 1 | 15 | |||
AK218349.1 | - | 1 | 15 | |||
AK219247.1 | - | 1 | 15 | |||
BC067064.1 | - | 7 | 1657 | 30537997 | ||
BC082330.1 | - | 7 | 1458 | 30539879 | ||
EU554632.1 | - | 7 | 1355 | |||
X63349.1 | - | 7 | 1759 |