Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110222.1 | NP_001103692.1 | 8 | 1748 | ||
NM_001110223.1 | NP_001103693.1 | 8 | 1670 | |||
NM_001110224.1 | NP_001103694.1 | 7 | 1667 | |||
NM_010025.2 | NP_034155.2 | 7 | 1652 | |||
GenBank mRNA | AB011678.1 | - | 8 | 1766 | ||
AK164119.1 | - | 7 | 1554 | |||
AY560329.1 | - | 8 | 1558 | |||
BC056391.1 | - | 7 | 1560 | 6855006 | ||
BC057010.1 | - | 7 | 1652 | 6825750 | ||
BC062974.1 | - | 7 | 1652 | 6825750 |