Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276461.1 | NP_001263390.1 | 5 | 585 | ||
NM_001276462.1 | NP_001263391.1 | 3 | 494 | |||
NM_001276463.1 | NP_001263392.1 | 3 | 494 | |||
NM_001276467.1 | NP_001263396.1 | 5 | 585 | |||
NM_010026.3 | NP_034156.2 | 5 | 585 | |||
GenBank mRNA | AF075461.1 | - | 5 | 404 | ||
AF075462.1 | - | 5 | 404 | |||
AK122477.1 | - | 3 | 189 | |||
AK132979.1 | - | 3 | 324 | |||
AK146545.1 | - | 5 | 423 | |||
AK147689.1 | - | 3 | 495 | |||
AK161586.1 | - | 495 | ||||
BC048818.1 | - | 3 | 460 | |||
BC094581.1 | - | 5 | 585 | 30549241 | ||
CB527210.1 | - | 3 | 288 | |||
CD355387.1 | - | 1 | 82 |