Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010046.2 | NP_034176.1 | 7 | 693 | ||
GenBank mRNA | AB057816.1 | - | 7 | 681 | ||
AF078752.1 | - | 7 | 751 | |||
BC003717.1 | - | 7 | 693 | 3486724 |